Alfa Aesar™ 5-Bromo-2-furaldehyde, 98%, stab. with 2% ethanol

Manufacturer: Alfa Aesar™ B2253618

Quantity 50g

 View more versions of this product

Catalog No. AAB2253618

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1899-24-7
    Molecular Formula C5H3BrO2
    Formula Weight 174.98
    MDL Number MFCD00159501
    Beilstein 108403
    Synonym 5-Bromofuran-2-carboxaldehyde; 5-Bromofurfural
    Chemical Name or Material 5-Bromo-2-furaldehyde, stabilized with 2% ethanol

    Specifications

    Formula Weight 174.98
    Molecular Formula C5H3BrO2
    CAS 1899-24-7
    Melting Point 82° to 85°C
    Quantity 50g
    MDL Number MFCD00159501
    Beilstein 108403
    Synonym 5-Bromofuran-2-carboxaldehyde; 5-Bromofurfural
    Boiling Point 112°C (6mmHg)
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 5-Bromo-2-furaldehyde, stabilized with 2% ethanol
    Assay 98%

    GHS H Statement
    H301-H315-H319-H335
    Toxic if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P301+P310-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit