Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-furaldehyde 98.0+%, TCI America™
SDP

Catalog No. B195425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B195425G 25 g
B19545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B195425G Supplier TCI America Supplier No. B195425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1899-24-7
Molecular Formula C5H3BrO2
Molecular Weight (g/mol) 174.98
MDL Number MFCD00159501
InChI Key WJTFHWXMITZNHS-UHFFFAOYSA-N
Synonym 5-bromo-2-furaldehyde, 5-bromofurfural, 5-bromo-2-furfural, 5-bromofuran-2-carboxaldehyde, 5-bromofurancarboxaldehyde, 2-furancarboxaldehyde, 5-bromo, 5-bromofuraldehyde, 5-bromo-furan-2-carbaldehyde, 2-furaldehyde, 5-bromo, 5-bromo-furfural
PubChem CID 600328
IUPAC Name 5-bromofuran-2-carbaldehyde
SMILES BrC1=CC=C(O1)C=O

Specifications

Melting Point 84°C
Boiling Point 112°C
Quantity 25 g
Formula Weight 174.98
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-furaldehyde
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.