Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Bromo-2'-hydroxyacetophenone 97.0+%, TCI America™
SDP

Catalog No. B206125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B206125G 25 g
B20615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B206125G Supplier TCI America Supplier No. B206125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1450-75-5
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.046
MDL Number MFCD00191850
InChI Key HQCCNFFIOWYINW-UHFFFAOYSA-N
Synonym 5'-bromo-2'-hydroxyacetophenone, 1-5-bromo-2-hydroxyphenyl ethanone, 2-acetyl-4-bromophenol, 5-bromo-2-hydroxyacetophenone, 2'-hydroxy-5'-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl ethan-1-one, ethanone, 1-5-bromo-2-hydroxyphenyl, 2-hydroxy-5-bromoacetophenone, 1-5-bromo-2-hydroxyphenyl-ethanon
PubChem CID 95991
IUPAC Name 1-(5-bromo-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Br)O

Specifications

Melting Point 58°C
Color White-Yellow
Quantity 25 g
Formula Weight 215.05
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Bromo-2′-hydroxyacetophenone
RTECSNumber KM5556350
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.