Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methoxyphenol 98.0+%, TCI America™
SDP

Catalog No. B334225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B334225G 25 g
B33425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B334225G Supplier TCI America Supplier No. B334225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 37942-01-1
Molecular Formula C7H7BrO2
Molecular Weight (g/mol) 203.035
MDL Number MFCD00673137
InChI Key OLSJHVZRUFFIPL-UHFFFAOYSA-N
Synonym 4-Bromo-2-hydroxyanisole
PubChem CID 142228
IUPAC Name 5-bromo-2-methoxyphenol
SMILES COC1=C(C=C(C=C1)Br)O

Specifications

Melting Point 65°C
Color White-Yellow
Boiling Point 150°C
Quantity 25 g
Formula Weight 203.04
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxyphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.