Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride) 98.0+%, TCI America™
SDP

Catalog No. B51731G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51731G 1 g
B51735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51731G Supplier TCI America Supplier No. B51731G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89694-45-1
Molecular Formula C7H8BBrO3
Molecular Weight (g/mol) 230.852
MDL Number MFCD00093409
InChI Key FVRLSHRAYPFXRQ-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxyphenyl boronic acid, 5-bromo-2-methoxybenzeneboronic acid, 4-bromoanisole-2-boronic acid, 3-bromo-6-methoxyphenylboronic acid, boronic acid,b-5-bromo-2-methoxyphenyl, 5-bromo-2-methoxyphenylboronicacid, rarechem ah pb 0045, acmc-209r2g, amtb789, 2-methoxy-5-bromophenylboronic acid
PubChem CID 4532730
IUPAC Name (5-bromo-2-methoxyphenyl)boronic acid
SMILES B(C1=C(C=CC(=C1)Br)OC)(O)O

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 1 g
Formula Weight 230.85
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxyphenylboronic Acid (contains varying amounts of Anhydride)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.