Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. B305025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B305025G 25 g
B30505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B305025G Supplier TCI America Supplier No. B305025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79669-49-1
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00267350
InChI Key SEENCYZQHCUTSB-UHFFFAOYSA-N
Synonym 2-methyl-5-bromobenzoic acid, 5-bromo-o-toluic acid, 5-bromo-2-methyl-benzoic acid, 5-bromo-2-methyl benzoic acid, synquest 2721-9-x8, benzoic acid, 5-bromo-2-methyl, pubchem4728, acmc-1bhwo, 3-bromo-6-methylbenzoic acid, ksc497q7f
PubChem CID 346004
IUPAC Name 5-bromo-2-methylbenzoic acid
SMILES CC1=CC=C(Br)C=C1C(O)=O

Specifications

Melting Point 171°C
Color White-Yellow
Quantity 25 g
Formula Weight 215.05
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.