Alfa Aesar™ 5-Bromo-2-phenyl-7-azaindole, 97%

Manufacturer: Alfa Aesar™ H3519303

Quantity 1g

 View more versions of this product

Catalog No. AAH3519303

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 953414-75-0
    Formula Weight 273.12
    MDL Number MFCD11109822
    Synonym 5-Bromo-2-phenyl-1H-pyrrolo[3,2-b]pyridine
    Chemical Name or Material 5-Bromo-2-phenyl-7-azaindole

    Specifications

    Melting Point 287° to 289°C
    Formula Weight 273.12
    Color Pale brown
    Physical Form Solid
    Sensitivity Light Sensitive
    MDL Number MFCD11109822
    Synonym 5-Bromo-2-phenyl-1H-pyrrolo[3,2-b]pyridine
    Quantity 1g
    CAS 953414-75-0
    Chemical Name or Material 5-Bromo-2-phenyl-7-azaindole
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit