Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-thiophenesulfonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. B287825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B287825G 25 g
B28785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B287825G Supplier TCI America Supplier No. B287825G

Chemical Identifiers

CAS 55854-46-1
Molecular Formula C4H2BrClO2S2
Molecular Weight (g/mol) 261.53
MDL Number MFCD00084909
InChI Key WGYBIEOLAFYDEC-UHFFFAOYSA-N
Synonym 5-bromothiophenesulfonyl chloride, 5-bromo-2-thiophenesulfonyl chloride, 5-bromo-thiophene-2-sulfonyl chloride, 5-bromothiophene-2-sulphonyl chloride, 5-bromo-2-chlorosulphonyl thiophene, 5-bromo-2-chlorosulfonyl thiophene, 5-bromothiophenesulphonyl chloride, buttpark 21\07-09, 5-bromo 2-thienyl chlorosulfone, 5-bromothiophen-2-sulfonyl chloride
PubChem CID 2733924
IUPAC Name 5-bromothiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(Br)S1

Specifications

Melting Point 41°C
Color Yellow
Boiling Point 110°C
Quantity 25 g
UN Number 3261
Formula Weight 261.53
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-thiophenesulfonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.