Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-3-chloro-2-fluoropyridine, 96%, Thermo Scientific Chemicals

Catalog No. AAH6454703
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6454703 1 g
AAH6454706 5 g
AAH64547MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6454703 Supplier Thermo Scientific Chemicals Supplier No. H6454703

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38185-56-7
Molecular Formula C5H2BrClFN
Molecular Weight (g/mol) 210.43
MDL Number MFCD09834365
InChI Key WXKLHBHJIBAXQH-UHFFFAOYSA-N
Synonym 2-fluoro-3-chloro-5-bromopyridine, 3-bromo-5-chloro-6-fluoropyridine, 5-bromo-3-chloro-2-fluoro-pyridine, acmc-1ct80, 5-bromo-3-chloro-2-fluoropyridine,, pyridine,5-bromo-3-chloro-2-fluoro, pyridine, 5-bromo-3-chloro-2-fluoro
PubChem CID 12056904
IUPAC Name 5-bromo-3-chloro-2-fluoropyridine
SMILES FC1=NC=C(Br)C=C1Cl

Specifications

Melting Point 60°C
Quantity 1 g
Beilstein 1563886
Formula Weight 210.43
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 5-Bromo-3-chloro-2-fluoropyridine
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.