Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide cyclohexylammonium salt hydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
25mg
100mg
Packaging:
Glass bottle
114162-64-0
C6H13N·xH2O
521.79
MFCD00058543
OMJGCRHKCXJGSX-ZSRZBWJVSA-N
5-bromo-4-chloro-3-indolyl,a-d-glucuronide cyclohe,cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate hydrate
126842357
(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium;hydrate
C1CCC(CC1)[NH3+].C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.O
114162-64-0
C6H13N·xH2O
521.79
MFCD00058543
OMJGCRHKCXJGSX-ZSRZBWJVSA-N
5-bromo-4-chloro-3-indolyl,a-d-glucuronide cyclohe,cyclohexylammonium ion 2s,3s,4s,5r,6s-6-5-bromo-4-chloro-1h-indol-3-yl oxy-3,4,5-trihydroxyoxane-2-carboxylate hydrate
126842357
(2S,3S,4S,5R,6S)-6-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-3,4,5-trihydroxyoxane-2-carboxylate;cyclohexylazanium;hydrate
C1CCC(CC1)[NH3+].C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)C(=O)[O-])O)O)O)Cl)Br.O
Melting Point 230°C
Water 5% max. (K.F.)
Packaging Glass bottle
Color Colorless or White
Quantity 100mg
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Solubility Information Solubility in water: soluble
Formula Weight 521.79
Physical Form Powder, Needles or Crystals
Percent Purity 98%
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-glucuronide cyclohexylammonium salt hydrate, 98%
Show More Show Less

MOISTURE SENSITIVE

LIGHT SENSITIVE

TSCA : TSCA