Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloro-3-indolyl Phosph., p-Toluidine Salt (Wht. to Off-Wht. Powd.), Fisher BioReagents

Catalog No. p-24848 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Amber Glass
BCIP is used in conjunction with Nitro Blue Tetrazolium (NBT) in the colorimetric detection of Alkaline Phosphatase. It is suitable for use in immunohistochemistry, immunoblot staining, and ELISA applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H15BrClN2O4P
Molecular Weight (g/mol) 433.62
InChI Key QEIFSLUFHRCVQL-UHFFFAOYSA-N

Specifications

Alpha Vector Bromo-4-chloro-3-indolyl Phosphate,p-Toluidine Salt (BCIP)
CAS 6578-06-9
Melting Point 222.8°C
Color White
Assay Percent Range ≥99 %
Packaging Amber Glass
Quantity 100 mg
Synonym BCIP
Formula Weight 433.62
Percent Purity 99% min.
Physical Form Solid
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl Phosphate, p-Toluidine Salt
Show More Show Less
Recommended Storage <0°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.