Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-4-chloro-3-indolyl phosphate disodium salt, 98%

Catalog No. aaj64451md
Click to view available options
Quantity:
250 mg
1 g

Substrate for alkaline phosphatase | CAS: 102185-33-1 | C8H4BrClNNa2O4P | 370.43 g/mol

5-Bromo-4-chloro-3-indolyl phosphate disodium salt, is used for the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-4-chloro-3-indolyl phosphate disodium salt, is used for the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

Solubility
Soluble in water (50 mg/mL). Insoluble in dimethylformamide.

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102185-33-1
Molecular Formula C8H4BrClNNa2O4P
Molecular Weight (g/mol) 370.43
MDL Number MFCD00036757
InChI Key OAZUOCJOEUNDEK-UHFFFAOYSA-L
PubChem CID 6097197
IUPAC Name disodium;(5-bromo-4-chloro-1H-indol-3-yl) phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12

Specifications

Color Blue to White
Quantity 250 mg
Sensitivity Light and moisture sensitive
Synonym sodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 5-bromo-4-chloro-3-indolyl phosphate disodium salt, 5-bromo-4-chloro-3-indoxyl phosphate, disodium salt, bcip™, 1h-indol-3-ol, 5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, disodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 1h-indol-3-ol,5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, 5-bromo-4-chloro-3-indolyl phosphate, 5-bromo-4-chloro-3-indolylphosphate disodium salt
Solubility Information Soluble in water (50mg/mL). Insoluble in dimethylformamide.
Formula Weight 370.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl phosphate disodium salt
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.