Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-6-chloro-3-indolyl beta-D-Galactopyranoside (contains ca. 10% Ethyl Acetate) 98.0+%, TCI America™
SDP

Catalog No. B3469100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3469100MG 100 mg
B346920MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3469100MG Supplier TCI America Supplier No. B3469100MG

[for Biochemical Research]

Chemical Identifiers

CAS 93863-88-8
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.63
MDL Number MFCD00216824
InChI Key CHRVKCMQIZYLNM-MBJXGIAVSA-N
Synonym 5-bromo-6-chloro-3-indolyl-beta-d-galactoside, 5-bromo-6-chloro-3-indolyl b-d-galactopyranoside, magenta-gal, 5-bromo-6-chloro-3-indolyl-beta-d-galactopyranoside, 2s,3r,4s,5r,6r-2-5-bromo-6-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, magenta tm-beta-d-gal, 5-bromo-6-chloro-3-indoxyl-beta-d-galactopyranoside, 5-bromo-6-chloro-3-indolyl beta-d-galactopyranoside, magenta tm-beta-d-galactoside, 5-bromo-6-chloro-3-indolyl-b-d-galactopyranoside
PubChem CID 2733803
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-6-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1O[C@@H](OC2=CNC3=CC(Cl)=C(Br)C=C23)[C@H](O)[C@@H](O)[C@H]1O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 408.63
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-6-chloro-3-indolyl beta-D-Galactopyranoside (contains ca. 10% Ethyl Acetate) [for Biochemica
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.