Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-8-hydroxyquinoline 96.0+%, TCI America™
SDP

Catalog No. B332425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B332425G 25 g
B33245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B332425G Supplier TCI America Supplier No. B332425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1198-14-7
Molecular Formula C9H6BrNO
Molecular Weight (g/mol) 224.057
MDL Number MFCD00457011
InChI Key WIIUANWSGSTCLG-UHFFFAOYSA-N
Synonym 5-Bromo-8-quinolinol
PubChem CID 70974
IUPAC Name 5-bromoquinolin-8-ol
SMILES C1=CC2=C(C=CC(=C2N=C1)O)Br

Specifications

Melting Point 127°C
Color White-Yellow
Quantity 25 g
Formula Weight 224.06
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-8-hydroxyquinoline
RTECSNumber VC4582000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.