Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Bromo-8-methoxyquinoline, 96%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
5-Bromo-8-methoxyquinoline is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Bromo-8-methoxyquinoline is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents and heat.
5-Bromo-8-methoxyquinoline is used as an organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Specifications
Melting Point | 80°C to 82°C |
Quantity | 250 mg |
Solubility Information | Slightly soluble in water. |
Formula Weight | 238.08 |
Percent Purity | 96% |
Chemical Name or Material | 5-Bromo-8-methoxyquinoline |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction