Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromoacenaphthene 93.0+%, TCI America™
SDP

Catalog No. B128325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B128325G 25 g
B12835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B128325G Supplier TCI America Supplier No. B128325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2051-98-1
Molecular Formula C12H9Br
Molecular Weight (g/mol) 233.11
MDL Number MFCD00003809
InChI Key QALKJGMGKYKMKE-UHFFFAOYSA-N
Synonym 5-bromoacenaphthene, acenaphthene, 5-bromo, acenaphthylene, 5-bromo-1,2-dihydro, 5-bromoacenaphtene, 5-bromo acenaphthene, akos 51, maybridge1_006415, acmc-1ccr7
PubChem CID 74923
IUPAC Name 5-bromo-1,2-dihydroacenaphthylene
SMILES BrC1=C2C=CC=C3CCC(C=C1)=C23

Specifications

Melting Point 55°C
Color Yellow
Boiling Point 335°C
Quantity 25 g
Formula Weight 233.11
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoacenaphthene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.