Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromobenzo[b]thiophene-2-carboxylic acid, ≥97%, Thermo Scientific™

Catalog No. CC38801DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC38801DA 1 g
CC38801CB 250 mg
CC38801DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC38801DA Supplier Thermo Scientific Chemicals Supplier No. CC38801DA
Add to Cart

Chemical Identifiers

CAS 7312-10-9
Molecular Formula C9H4BrO2S
Molecular Weight (g/mol) 256.09
MDL Number MFCD01929338
InChI Key ONNFNEFYXIPHCA-UHFFFAOYSA-M
Synonym 5-bromo-1-benzothiophene-2-carboxylic acid, 5-bromobenzo b thiophene-2-carboxylic acid, 5-bromo-benzo b thiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 5-bromo, onnfnefyxiphca-uhfffaoysa-n, 5-bromobenzothiophene-2-carboxylic acid, benzo b thiophene-2-carboxylicacid, 5-bromo, 5-bromo-1-benzo b thiophene-2-carboxylic acid
PubChem CID 737737

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 257.11
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-benzothiophene-2-carboxylic acid
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.