Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromopentyl acetate, 98%

Catalog No. AAA1554914
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 15848-22-3 | C7H13BrO2 | 209.08 g/mol

5-?Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-​Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15848-22-3
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00039204
InChI Key JIHJLQWUVZUKCH-UHFFFAOYSA-N
Synonym 5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester
PubChem CID 85140
IUPAC Name 5-bromopentyl acetate
SMILES CC(=O)OCCCCCBr

Specifications

Density 1.255
Boiling Point 109°C to 110°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.462
Quantity 25 g
Beilstein 1752109
Solubility Information Slightly soluble in water.
Formula Weight 209.09
Percent Purity 98%
Chemical Name or Material 5-Bromopentyl acetate
EINECSNumber 239-961-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.