Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromopyridine-2-carboxylic acid, 98%

Catalog No. p-7028386
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2567506

Supplier: Thermo Scientific Chemicals B2567506

Only null left
Add to Cart

CAS: 30766-11-1 | C6H3BrNO2 | 201.00 g/mol

5-Bromopyridine-2-carboxylic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromopyridine-2-carboxylic acid is used as a pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30766-11-1
Molecular Formula C6H3BrNO2
Molecular Weight (g/mol) 201.00
MDL Number MFCD00234149
InChI Key MNNQIBXLAHVDDL-UHFFFAOYSA-M
Synonym 5-bromopicolinic acid, 5-bromo-2-pyridinecarboxylic acid, 2-pyridinecarboxylic acid, 5-bromo, 5-bromo-2-carboxypyridine, 5-bromo-pyridine-2-carboxylic acid, 5-bromo-2-picolinic acid, 5-bromo-2-carboxy pyridine, 5-bromo-2-pyridine carboxylic acid, 5-bromopyridine-2-carboxylicacid
PubChem CID 608544
IUPAC Name 5-bromopyridine-2-carboxylic acid
SMILES [O-]C(=O)C1=CC=C(Br)C=N1

Specifications

Melting Point 173°C to 175°C
Quantity 5 g
Solubility Information Soluble in methanol.
Formula Weight 202.01
Percent Purity 98%
Chemical Name or Material 5-Bromopyridine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.