Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromosalicylaldehyde, 99%

Catalog No. p-77412
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 1761-61-1 | C7H5BrO2 | 201.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1761-61-1
Molecular Formula C7H5BrO2
Molecular Weight (g/mol) 201.02
MDL Number MFCD00003330
InChI Key MKKSTJKBKNCMRV-UHFFFAOYSA-N
Synonym 5-bromosalicylaldehyde, benzaldehyde, 5-bromo-2-hydroxy, 2-hydroxy-5-bromobenzaldehyde, salicylaldehyde, 5-bromo, 5-bromo-salicyclaldehyde, unii-0z65a0a8ce, 5-bromo-2-hydroxy-benzaldehyde, 5-bromo-salicyclic aldehyde, 5-bromo-2-hydroxy benzaldehyde, pubchem3779
PubChem CID 72863
IUPAC Name 5-bromo-2-hydroxybenzaldehyde
SMILES C1=CC(=C(C=C1Br)C=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 103°C to 107°C
Color Beige-Yellow to Yellow
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula BrC6H3(OH)CHO
Quantity 25 g
Beilstein 08, 54
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 201.02
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 5-Bromosalicylaldehyde, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 217-167-2
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.