Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromothiazole, 97%

Catalog No. AAH5538703
Click to view available options
Quantity:
1 g

CAS: 3034-55-7 | C3H2BrNS | 164.02 g/mol

5-Bromothiazole is used as a pharmaceutical intermediate and performance material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromothiazole is used as a pharmaceutical intermediate and performance material.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-55-7
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.02
MDL Number MFCD07787394
InChI Key DWUPYMSVAPQXMS-UHFFFAOYSA-N
PubChem CID 546059
IUPAC Name 5-bromo-1,3-thiazole
SMILES C1=C(SC=N1)Br

Specifications

Density 1.835
Flash Point 76°C (169°F)
Refractive Index 1.5955
Quantity 1 g
Sensitivity Moisture sensitive
Synonym 5-bromothiazole, thiazole, 5-bromo, 5-bromo-thiazole, 5-bromthiazole, thiazole,5-bromo, 5-bromothiazol, 5-bromo thiazole, zlchem 497, pubchem2077, 2-bromo-1,4-thiazole
Solubility Information Soluble in water.
Formula Weight 164.02
Percent Purity 98%
Chemical Name or Material 5-Bromothiazole
Hazard Category H227-H302-H315-H317-H318-H335
Hazard Statement GHS H Statement
H301-H318-H227-H315-H317-H335
Toxic if swallowed.
Causes serious eye damage.
Combustible liquid.
Causes skin irritation.
May cause an allergic skin reaction.
May cause respiratory irritation.
Precautionary Statement P210-P235-P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P370+P378q-P501c
TSCA No
Recommended Storage Store at -20°C; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.