Alfa Aesar™ 5-Bromovaleric acid, 97%

Manufacturer: Alfa Aesar™ A1176530

Quantity 250g

 View more versions of this product

Catalog No. AAA1176530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2067-33-6
    Molecular Formula Br(CH2)4CO2H
    Formula Weight 181.03
    MDL Number MFCD00004414
    UN Number UN3261
    Beilstein 1745664
    Synonym 5-Bromopentanoic acid
    Chemical Name or Material 5-Bromovaleric acid

    Specifications

    Formula Weight 181.03
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 142° to 145°C (13mmHg)
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Molecular Formula Br(CH2)4CO2H
    Beilstein 1745664
    Synonym 5-Bromopentanoic acid
    Melting Point 38° to 41°C
    TSCA TSCA
    MDL Number MFCD00004414
    UN Number UN3261
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    CAS 2067-33-6
    Quantity 250g
    Flash Point >100°C (212°F)
    Chemical Name or Material 5-Bromovaleric acid
    Assay 97%

    Description

    Applications
    5-Bromovaleric acid is used in the synthesis of inhibitors for aminoglycoside resistant bacteria. 5-Bromovaleric acid is also used in the preparation of sequence specific base pair mimics as topoisomerase IB inhibitors.

    Solubility
    Soluble in ethanol. Insoluble in water.

    Notes
    Store away from strong bases and oxidizing agents. Store at room temperature.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF SWALLOWED: Rinse mouth.
    Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit