Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Carboxylcytosine (5-caC) Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$147.60 - $584.25
Specifications
Antigen | 5-Carboxylcytosine (5-caC) |
---|---|
Concentration | 1 mg/mL |
Applications | Dot Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3251
![]() ![]() |
Active Motif
61230 |
10 μg |
Each for $147.60
|
|
|||||
50-199-3250
![]() ![]() |
Active Motif
61229 |
100 μg |
Each for $584.25
|
|
|||||
Description
5-carboxylcytosine is the final oxidized derivative of 5-methylcytosine, a type of DNA methylation. 5-methylcytosine is oxidized to 5-hydroxymethylcytosine which is then oxidized to 5-formylcytosine and later to 5-carboxylcytosine. These oxidations are catalyzed by the Ten-Eleven Translocation (TET) enzyme family. 5-carboxylcytosine has been shown to cause reduced fidelity of RNA Polymerase II. Decreased levels of 5-hydroxymethylcytosine has been found in some cancers.Specifications
5-Carboxylcytosine (5-caC) | |
Dot Blot | |
Unconjugated | |
Rabbit | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
IgG | |
Protein A | |
Antibody |
1 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
This 5-Carboxylcytosine antibody was raised against 5-carboxylcytidine conjugated to kLH and recognizes 5-carboxylcytosine. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title