Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-chloro-1,3-dimethoxybenzene, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
Glass bottle,Glass Bottle
7051-16-3
C8H9ClO2
172.608
MFCD00008382
WQHNWJBSROXROL-UHFFFAOYSA-N
5-chloro-1,3-dimethoxybenzene,3,5-dimethoxychlorobenzene,benzene, 1-chloro-3,5-dimethoxy,5-chlororesorcinol dimethyl ether,1,3-dimethoxy-5-chlorobenzene,pubchem3042,benzene,5-dimethoxy,acmc-209oev,wln: gr co1 eo1,3,5-dimethoxyphenyl chloride
81502
1-chloro-3,5-dimethoxybenzene
COC1=CC(=CC(=C1)Cl)OC
7051-16-3
C8H9ClO2
172.608
MFCD00008382
WQHNWJBSROXROL-UHFFFAOYSA-N
5-chloro-1,3-dimethoxybenzene,3,5-dimethoxychlorobenzene,benzene, 1-chloro-3,5-dimethoxy,5-chlororesorcinol dimethyl ether,1,3-dimethoxy-5-chlorobenzene,pubchem3042,benzene,5-dimethoxy,acmc-209oev,wln: gr co1 eo1,3,5-dimethoxyphenyl chloride
81502
1-chloro-3,5-dimethoxybenzene
COC1=CC(=CC(=C1)Cl)OC
Melting Point 33.0°C to 37.0°C
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle,Glass Bottle
Color White to Yellow or Pink
Flash Point 112°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula ClC6H3(OCH3)2
Beilstein 06, III, 4335
Formula Weight 172.61
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 5-Chloro-1, 3-dimethoxybenzene
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.