Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-furaldehyde, 97%, Thermo Scientific™

Catalog No. ac394650010
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 21508-19-0
Molecular Formula C5H3ClO2
Molecular Weight (g/mol) 130.527
MDL Number MFCD02752599
InChI Key DGAUAVDWXYXXGQ-UHFFFAOYSA-N
Synonym 5-chloro-2-furaldehyde, 5-chlorofurfural, 2-furancarboxaldehyde, 5-chloro, 5-chloro-furfural, 2-chlorofuran-5-aldehyde, acmc-1cq7z, 5-chloro-furan-2carbaldehyde, 5-chlorofuran-2 carbaldehyde, 5-chloro-furan-2-carbaldehyde, 5-chlorofuran-2-carboxaldehyde
PubChem CID 2769630
IUPAC Name 5-chlorofuran-2-carbaldehyde
SMILES C1=C(OC(=C1)Cl)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Stabilizer 1% Ethanol
Formula Weight 130.53
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-furaldehyde, Stabilized
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if inhaled.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.