Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-iodopyrimidine, 98%, Thermo Scientific™

Catalog No. AAH6418506
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6418506 5 g
AAH64185MD 250 mg
AAH6418503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6418506 Supplier Thermo Scientific Chemicals Supplier No. H6418506

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Useful Intermediate for organic synthesis and pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store in dark. Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874676-81-0
Molecular Formula C4H2ClIN2
Molecular Weight (g/mol) 240.428
MDL Number MFCD08062390
InChI Key BWDLBDLGVMLSCS-UHFFFAOYSA-N
Synonym pyrimidine, 5-chloro-2-iodo, pubchem14957, acmc-209qnq, 5-chloro-2-iodo pyrimidine, ksc493q5d
PubChem CID 29938005
IUPAC Name 5-chloro-2-iodopyrimidine
SMILES C1=C(C=NC(=N1)I)Cl

Specifications

Color White
Quantity 5 g
Beilstein 14516398
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 240.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Chloro-2-iodopyrimidine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.