Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methylphenol, 98%

Catalog No. AAB2552706
Click to view available options
Quantity:
5 g
25 g

CAS: 5306-98-9 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5306-98-9
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00060672
InChI Key KKFPXGXMSBBNJI-UHFFFAOYSA-N
Synonym 5-chloro-o-cresol, 5-chloro-2-cresol, o-cresol, 5-chloro, phenol, 5-chloro-2-methyl, 3-chloro-6-methylphenol, 2-methyl-5-chlorophenol, 4-chloro-2-hydroxytoluene, 5-chloro-2-methyl-phenol, phenol,5-chloro-2-methyl
PubChem CID 79192
IUPAC Name 5-chloro-2-methylphenol
SMILES CC1=CC=C(Cl)C=C1O

Specifications

Melting Point 70°C to 75°C
Boiling Point 109°C to 112°C (15 mmHg)
Quantity 5 g
UN Number UN3437
Beilstein 1906683
Formula Weight 142.59
Percent Purity 98%
Chemical Name or Material 5-Chloro-2-methylphenol
Hazard Category H302+H312+H332-H315-H318-H335
Hazard Statement GHS H Statement
H301-H311-H331-H318-H315-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROCRESOLS, SOLID
EINECSNumber 226-160-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.