Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-nitroaniline 99.0+%, TCI America™
SDP

Catalog No. C1139250G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g
500 g

Chemical Identifiers

CAS 1635-61-6
Molecular Formula C6H5ClN2O2
Molecular Weight (g/mol) 172.568
MDL Number MFCD00007776
InChI Key ZCWXYZBQDNFULS-UHFFFAOYSA-N
Synonym 2-nitro-5-chloroaniline, benzenamine, 5-chloro-2-nitro, 3-chloro-6-nitroaniline, 5-chloro-2-nitrobenzeneamine, aniline, 5-chloro-2-nitro, 2-amino-4-chloro-nitrobenzene, unii-656xjm4con, 5-chloro-2-nitrobenzenamine, ccris 8934, 5-chloro-2-nitro aniline
PubChem CID 74218
IUPAC Name 5-chloro-2-nitroaniline
SMILES C1=CC(=C(C=C1Cl)N)[N+](=O)[O-]

Specifications

Melting Point 128°C
Quantity 250 g
UN Number 2237
Formula Weight 172.57
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitroaniline
EINECSNumber (3)-0407
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.