Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-nitroanisole 97%, Thermo Scientific™

Catalog No. AC109590100
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC109590100 10 g
AC109590250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC109590100 Supplier Thermo Scientific Chemicals Supplier No. 109590100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6627-53-8
Molecular Formula C7H6ClNO3
Molecular Weight (g/mol) 187.579
InChI Key ABEUJUYEUCCZQF-UHFFFAOYSA-N
Synonym 5-chloro-2-nitroanisole, 4-chloro-2-methoxynitrobenzene, 2-nitro-5-chloroanisole, benzene, 4-chloro-2-methoxy-1-nitro, anisole, 2-nitro-5-chloro, anisole, 5-chloro-2-nitro, 5-chloro-2-nitro-1-methoxybenzene, 4-chloro-2-methoxy-1-nitro-benzene, pubchem4104, acmc-1badb
PubChem CID 81110
IUPAC Name 4-chloro-2-methoxy-1-nitrobenzene
SMILES COC1=C(C=CC(=C1)Cl)[N+](=O)[O-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Green to Yellow
Infrared Spectrum Authentic
Linear Formula ClC6H3(NO2)OCH3
Quantity 10 g
Beilstein 06, 239
Formula Weight 187.58
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitroanisole, 97%
Show More Show Less
EINECSNumber 229-594-1
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.