Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-3-ethyl-2-methylbenzothiazolium p-toluenesulfonate (Pract), 95%, Thermo Scientific™

Catalog No. AC404620250
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63149-16-6
Molecular Formula C17H18ClNO3S2
Molecular Weight (g/mol) 383.905
MDL Number MFCD00143454
InChI Key GYXTWUJZURSNCF-UHFFFAOYSA-M
Synonym 5-chloro-3-ethyl-2-methylbenzothiazolium p-toluenesulfonate, 5-chloro-3-ethyl-2-methylbenzo d thiazol-3-ium 4-methylbenzenesulfonate, benzothiazolium, 5-chloro-3-ethyl-2-methyl-, salt with 4-methylbenzenesulfonic acid 1:1, 5-chloro-3-ethyl-2-methylbenzothiazolium p-toluenesulphonate, benzothiazolium, 5-chloro-3-ethyl-2-methyl-, 4-methylbenzenesulfonate 1:1, 5-chloro-3-ethyl-2-methyl-1,3-benzothiazol-3-ium tosylate, 5-chloro-3-ethyl-2-methyl-1,3-benzothiazol-3-ium 4-methylbenzenesulfonate, 5-chloro-3-ethyl-2-methyl-1,3-benzothiazol-3-ium; 4-methylbenzenesulfonate
PubChem CID 113090
IUPAC Name 5-chloro-3-ethyl-2-methyl-1,3-benzothiazol-3-ium;4-methylbenzenesulfonate
SMILES CC[N+]1=C(SC2=C1C=C(C=C2)Cl)C.CC1=CC=C(C=C1)S(=O)(=O)[O-]

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Brown
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 25 g
Formula Weight 383.91
Percent Purity ≥94%
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-3-ethyl-2-methylbenzothiazolium p-toluenesulfonate, (Pract), 95%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.