Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorobenzimidazole 98.0+%, TCI America™
SDP

Catalog No. C013025G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4887-82-5
Molecular Formula C7H5ClN2
Molecular Weight (g/mol) 152.581
MDL Number MFCD00059698
InChI Key NKLOLMQJDLMZRE-UHFFFAOYSA-N
PubChem CID 78599
IUPAC Name 6-chloro-1H-benzimidazole
SMILES C1=CC2=C(C=C1Cl)NC=N2

Specifications

Melting Point 131°C
Quantity 25 g
Formula Weight 152.58
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorobenzimidazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.