Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorobenzo[b]thiophene, 97%, Thermo Scientific Chemicals

Catalog No. AAH6174814
Click to view available options
Quantity:
5 g
25 g

5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20532-33-6
Molecular Formula C8H5ClS
Molecular Weight (g/mol) 168.638
MDL Number MFCD00055623
InChI Key SNYURIHMNFPQFL-UHFFFAOYSA-N
Synonym 5-chlorobenzothiophene, 5-chlorobenzo b thiophene, 5-chloro-benzo b thiophene, 5-chloro benzothiophene, benzo b thiophene, 5-chloro, zlchem 66, 5-chlorothionaphthene, 5-chloranyl-1-benzothiophene, benzo b thiophene,5-chloro, ksc544i5d
PubChem CID 11309754
IUPAC Name 5-chloro-1-benzothiophene
SMILES C1=CC2=C(C=CS2)C=C1Cl

Specifications

Color White
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 168.64
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Chlorobenzo[b]thiophene
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.