Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloroindole-2-carboxylic Acid 98.0+%, TCI America™
SDP

Catalog No. C186825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C186825G 25 g
C18685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C186825G Supplier TCI America Supplier No. C186825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10517-21-2
Molecular Formula C9H5ClNO2
Molecular Weight (g/mol) 194.59
MDL Number MFCD00005613
InChI Key FUQOTYRCMBZFOL-UHFFFAOYSA-M
Synonym 5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783
PubChem CID 82693
IUPAC Name 5-chloro-1H-indole-2-carboxylate
SMILES [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1

Specifications

Melting Point 290°C
Color White-Yellow
Quantity 25 g
Formula Weight 195.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloroindole-2-carboxylic Acid
RTECSNumber NL5998400
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.