Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorosalicylanilide 98.0+%, TCI America™
SDP

Catalog No. C0286250G
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 4638-48-6
Molecular Formula C13H10ClNO2
Molecular Weight (g/mol) 247.678
MDL Number MFCD00002320
InChI Key KGYNGVVNFRUOOZ-UHFFFAOYSA-N
PubChem CID 14869
IUPAC Name 5-chloro-2-hydroxy-N-phenylbenzamide
SMILES C1=CC=C(C=C1)NC(=O)C2=C(C=CC(=C2)Cl)O

Specifications

Melting Point 206°C
Color White
Quantity 250 g
Formula Weight 247.68
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorosalicylanilide
RTECSNumber CV2450475
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.