Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorothiophene-2-carboxylic Acid 95.0+%, TCI America™
SDP

Catalog No. C123025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C123025G 25 g
C12305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C123025G Supplier TCI America Supplier No. C123025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24065-33-6
Molecular Formula C5H3ClO2S
Molecular Weight (g/mol) 162.59
MDL Number MFCD00041426
InChI Key QZLSBOVWPHXCLT-UHFFFAOYSA-N
Synonym 5-chloro-2-thiophenecarboxylic acid, 2-thiophenecarboxylic acid, 5-chloro, 2-chloro-5-carboxythiophene, 5-chloro-2-thiophene carboxylic acid, pubchem7380, maybridge3_003717, acmc-209g8i, rivaroxaban related compound, 5-18-06-00177 beilstein handbook reference, ksc204m2j
PubChem CID 95048
IUPAC Name 5-chlorothiophene-2-carboxylic acid
SMILES OC(=O)C1=CC=C(Cl)S1

Specifications

Melting Point 151°C
Quantity 25 g
Formula Weight 162.59
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorothiophene-2-carboxylic Acid
RTECSNumber XM8400000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.