Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorotryptamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. c11455g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 942-26-7
Molecular Formula C10H12Cl2N2
Molecular Weight (g/mol) 231.12
MDL Number MFCD00051990
InChI Key PBANXRNIXGEHPZ-UHFFFAOYSA-N
Synonym 5-chlorotryptamine hydrochloride, 2-5-chloro-1h-indol-3-yl ethanamine hydrochloride, 5-chlorotryptamine hcl, 5-chloro-1h-indole-3-ethylamine hydrochloride, 5-chloro-1h-indole-3-ethylamine monohydrochloride, 5-chlorotryptamine monohydrochloride, 2-5-chloro-1h-indol-3-yl ethylamine hydrochloride, 5-chloro-1h-indole-3-ethylamine hcl, 2-5-chloro-1h-indol-3-yl ethan-1-amine hydrochloride, 5-chloro-3-2-aminoethyl indole hydrochloride
PubChem CID 2827494
IUPAC Name 2-(5-chloro-1H-indol-3-yl)ethanamine;hydrochloride
SMILES C1=CC2=C(C=C1Cl)C(=CN2)CCN.Cl

Specifications

Quantity 5 g
Formula Weight 231.12
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorotryptamine Hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.