Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorovaleric Acid 96.0+%, TCI America™
SDP

Catalog No. C030925G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 1119-46-6
Molecular Formula C5H9ClO2
Molecular Weight (g/mol) 136.58
MDL Number MFCD00004415
InChI Key YSXDKDWNIPOSMF-UHFFFAOYSA-N
Synonym 5-Chloropentanoic Acid
PubChem CID 14244
ChEBI CHEBI:60701
IUPAC Name 5-chloropentanoic acid
SMILES OC(=O)CCCCCl

Specifications

Color Yellow
Boiling Point 136°C
Quantity 25 g
Formula Weight 136.58
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Chlorovaleric Acid
EINECSNumber (2)-1180
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.