Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyano-2-(trifluoromethyl)pyridine 98.0+%, TCI America™
SDP

Catalog No. C26985G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Catalog No. C26985G

Supplier: TCI America C26985G

Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 216431-85-5
Molecular Formula C7H3F3N2
Molecular Weight (g/mol) 172.11
MDL Number MFCD03001347
InChI Key ZBLULFJVRGVQAW-UHFFFAOYSA-N
Synonym 6-trifluoromethyl nicotinonitrile, 5-cyano-2-trifluoromethyl pyridine, 6-trifluoromethyl pyridine-3-carbonitrile, 6-trifluoromethyl-nicotinonitrile, 3-cyano-6-trifluoromethylpyridine, 3-pyridinecarbonitrile, 6-trifluoromethyl, 2-trifluoromethyl-5-cyanopyridine, pubchem14525, acmc-209fmx, ksc495e8b
PubChem CID 2777554
IUPAC Name 6-(trifluoromethyl)pyridine-3-carbonitrile
SMILES C1=CC(=NC=C1C#N)C(F)(F)F

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 172.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Cyano-2-(trifluoromethyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.