Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[5]Cycloparaphenylene 95.0+%, TCI America™
SDP

Catalog No. C2931100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
20 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2931100MG 100 mg
C293120MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2931100MG Supplier TCI America Supplier No. C2931100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96100-94-6
Molecular Formula C30H20
Molecular Weight (g/mol) 380.49
InChI Key JPDIIJOQNLNRQS-UHFFFAOYSA-N
Synonym [5]CPP
PubChem CID 131637028
SMILES C1CC2=C=C=C1C3=C=C=C(CC3)C4=C=C=C(CC4)C5=CC=C(C=C5)C6=C=C=C2CC6

Specifications

Quantity 100 mg
Formula Weight 380.49
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material [5]Cycloparaphenylene
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.