Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Decyne 98.0+%, TCI America™
SDP

Catalog No. D119025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D119025ML 25 mL
D11905ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D119025ML Supplier TCI America Supplier No. D119025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1942-46-7
Molecular Formula C10H18
Molecular Weight (g/mol) 138.254
MDL Number MFCD00027277
InChI Key JWBQJUFCNOLNNC-UHFFFAOYSA-N
Synonym 5-decyne, dibutylacetylene, 1,2-dibutylacetylene, acmc-209ey1, 5-decyne 8ci 9ci, 3,3?-diheptyloxacarbocyanine iodide
PubChem CID 16030
IUPAC Name dec-5-yne
SMILES CCCCC#CCCCC

Specifications

Color Yellow
Boiling Point 177°C
Quantity 25 mL
UN Number 3295
Formula Weight 138.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Decyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.