Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5'-Deoxy-5-fluorocytidine, Thermo Scientific™
$40.65 - $106.50
Specifications
Molecular Weight (g/mol) | 245.21 |
---|---|
Purity | ≥94 % |
Solubility Information | Miscible with methanol, DMSO |
Color | White to almost white |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AC468740010
|
Thermo Scientific Chemicals
468740010 |
1 g |
Each for $40.65
Order in Bulk
Request bulk or custom formats |
|
|||||
AC468740050
|
Thermo Scientific Chemicals
468740050 |
5 g |
Each for $106.50
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Specifications
245.21 | |
≥94 % | |
Miscible with methanol, DMSO | |
White to almost white |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title