Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Ethyl-m-xylene 98.0+%, TCI America™
SDP

Catalog No. e028201ml Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
0.1 mL

Chemical Identifiers

CAS 934-74-7
Molecular Formula C10H14
Molecular Weight (g/mol) 134.22
MDL Number MFCD00059235
InChI Key LMAUULKNZLEMGN-UHFFFAOYSA-N
Synonym 5-ethyl-m-xylene, 1,3-dimethyl-5-ethylbenzene, m-xylene, 5-ethyl, benzene, 1-ethyl-3,5-dimethyl, 5-ethyl-1,3-dimethylbenzene, benzene,5-dimethyl, 3,5-dimethylethylbenzene, acmc-209rm2, 1,3-dimethyl-5-ethyl benzene, 1-ethyl-3,5-dimethyl-benzene
PubChem CID 13627
IUPAC Name 1-ethyl-3,5-dimethylbenzene
SMILES CCC1=CC(C)=CC(C)=C1

Specifications

Melting Point -84°C
Color Yellow
Boiling Point 184°C
Quantity 0.1 mL
Physical Form Clear Liquid at 20°C
UN Number 3295
Formula Weight 134.22
Percent Purity ≥98.0% (GC)
Chemical Name or Material 5-Ethyl-m-xylene
EINECSNumber (3)-3427
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.