Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 5-Ethynyl-2'-deoxyuridine
GREENER_CHOICESDP

Catalog No. 720750 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
72-075-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 72-075-0 Supplier Tocris Bioscience™ Supplier No. 7207/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thymidine analog for labeling DNA; can be linked to a fluorophore by click chemistry

5-Ethynyl-2'-deoxyuridine is a thymidine analog for labeling DNA. Can be linked to an azide fluorophore through click chemistry and used to detect DNA synthesis in proliferating cells in vivo and in vitro. Detects very low levels of cell proliferation in tissues with low cellular turnover. 5-Ethynyl-2'-deoxyuridine suppresses cell proliferation in osteosarcoma and glioblastoma cell lines, induces a 'futile' DNA repair cycle and induces DNA strand breaks and apoptosis at a higher rate than Temozolomide (Cat. No. 2706).

Specifications

Chemical Name or Material 5-ethynyl-1-((2 R,4 S,5 R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1 H,3 H)-dione
CAS 61135-33-9
Quantity 50 mg
Target DNA, RNA and Protein Synthesis Compounds
Molecular Formula C11H12N2O5
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.