Learn More
Invitrogen™ EU (5-Ethynyl Uridine)

Description
Detect global RNA transcription temporally and spatially in both in-vitro and in-vivo with the modified nucleotide, ethynyl uridine. Approach uses powerful click chemistry detection, a simple and robust two-step technique. In step one, the alkyne-containing biomolecule is fed to cells or animals and actively incorporated into RNA. Detection utilizes the chemoselective ligation or 'click' reaction between an azide and an alkyne where the modified protein is detected with a corresponding azide-containing dye or hapten using the Click-iT Cell Reaction Buffer Kit. Cells can be analyzed by fluorescence microscopy, flow cytometry or high-content imaging and analysis (HCS).
Cell Analysis, Click Chemistries, Labeling Chemistry, Metabolic Labeling and Nascent Protein Synthesis with Click Chemistry
Order Info
Shipping Condition: Room temperature

Specifications
Specifications
Product Type | EU (5-Ethynyl Uridine) |
Content And Storage | Store at ≤-20°C |
Labeling Method | RNA Labeling |
Shipping Condition | Room Temperature |
Label or Dye | EU |
Quantity | 5 mg |
Product Line | Click-iT™, Molecular Probes™ |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.