Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals (+)-5-Fluoro-2'-deoxyuridine, 99+%

Catalog No. p-4510277
Click to view available options
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle

CAS: 50-91-9 | C9H11FN2O5 | 246.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-91-9
Molecular Formula C9H11FN2O5
Molecular Weight (g/mol) 246.19
MDL Number MFCD00006530
InChI Key ODKNJVUHOIMIIZ-RRKCRQDMSA-N
Synonym floxuridine, 2'-deoxy-5-fluorouridine, 5-fluoro-2'-deoxyuridine, 5-fluorodeoxyuridine, fluorodeoxyuridine, floxuridin, fudr, deoxyfluorouridine, fluoruridine deoxyribose, fdurd
PubChem CID 5790
ChEBI CHEBI:60761
IUPAC Name 5-fluoro-1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(F)C(=O)NC1=O

Specifications

Melting Point 148.0°C to 153.0°C
Color White
Loss on Drying 0.5% (60°C, 4 hrs) (vacuum) max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15,4144
Specific Rotation Condition + 35.90 (20.00°C c=1,water)
Specific Rotation + 35.90
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,practically insoluble in chcl3,insoluble in ether and hexane
Formula Weight 246.19
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material (+)-5-Fluoro-2′-deoxyuridine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.
IF ON SKIN: Wash with plenty of soap and
EINECSNumber 200-072-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.