Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl Violet, 80%, Thermo Scientific™
Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Dye Content | 75% min. |
Solubility Information | Solubility in water: slightly soluble. Other solubilities: soluble in methanol and ethanol |
Infrared Spectrum | Authentic |
Lambda Max | 593 to 599nm (c=0.1, H2O) |
Formula Weight | 492.14 |
Loss on Drying | 4% max. |
Percent Purity | 80% |
Quantity | 5 g |
Chemical Name or Material | Ethyl Violet |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction