Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-3-methylindole 98.0+%, TCI America™
SDP

Catalog No. F10531G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F10531G 1 g
F1053200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F10531G Supplier TCI America Supplier No. F10531G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 392-13-2
Molecular Formula C9H8FN
Molecular Weight (g/mol) 149.17
MDL Number MFCD03095377
InChI Key CSDHAGJNOQIBHZ-UHFFFAOYSA-N
PubChem CID 22351503
IUPAC Name 5-fluoro-3-methyl-1H-indole
SMILES CC1=CNC2=CC=C(F)C=C12

Specifications

Melting Point 85°C
Quantity 1 g
Formula Weight 149.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoro-3-methylindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.