Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoro-o-tolyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. F051525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F051525G 25 g
F05155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F051525G Supplier TCI America Supplier No. F051525G

Chemical Identifiers

CAS 175205-39-7
Molecular Formula C8H6FNS
Molecular Weight (g/mol) 167.201
InChI Key QVCYTKXGZLOVFA-UHFFFAOYSA-N
Synonym 5-fluoro-2-methylphenyl isothiocyanate, 2-methyl-5-fluorophenyl isothiocyanate, 5-fluoro-o-tolyl isothiocyanate, 5-fluoro-2-methylphenylisothiocyanate, isothiocyanic acid 5-fluoro-o-tolyl ester, acmc-20am3d, 4-fluoro-2-isothiocyanatotoluene, 2-methyl-5-fluorophenylisothiocyanate, 5-fluoro-2-methylbenzenisothiocyanate, 5-fluoro-2-methylphenyl-isothiocyanate
PubChem CID 519443
IUPAC Name 4-fluoro-2-isothiocyanato-1-methylbenzene
SMILES CC1=C(C=C(C=C1)F)N=C=S

Specifications

Color Yellow
Quantity 25 g
Formula Weight 167.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 5-Fluoro-o-tolyl Isothiocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.