Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoroindole, 98%

Catalog No. p-134748
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

Catalog No. AC311920010

Supplier: Thermo Scientific Chemicals 311920010

Only null left
Add to Cart

CAS: 399-52-0 | C8H6FN | 135.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 399-52-0
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00005671
InChI Key ODFFPRGJZRXNHZ-UHFFFAOYSA-N
Synonym 5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
PubChem CID 67861
ChEBI CHEBI:72818
IUPAC Name 5-fluoro-1H-indole
SMILES FC1=CC=C2NC=CC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 44.0°C to 48.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 135.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Fluoroindole, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 206-917-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.