Alfa Aesar™ 5-Fluoroindoline, 97%

Manufacturer: Alfa Aesar™ H54158MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH54158MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2343-22-8
    Molecular Formula C8H8FN
    Formula Weight 137.16
    MDL Number MFCD00214461
    Synonym 5-Fluoro-2,3-dihydroindole
    Chemical Name or Material 5-Fluoroindoline

    Specifications

    Formula Weight 137.16
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 250mg
    Molecular Formula C8H8FN
    MDL Number MFCD00214461
    CAS 2343-22-8
    Synonym 5-Fluoro-2,3-dihydroindole
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage 2° to 8°C
    Chemical Name or Material 5-Fluoroindoline
    Assay 97%

    Description

    Applications
    5-Fluoroindoline is used as pharmaceutical intermediate.

    Solubility
    Slightly soluble in water.

    Notes
    Air sensitive. Incompatible with oxidizing agents and heat.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2° to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit